siRNA Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246051.2 | XP_006246113.1 | 19 | 4128 | ||
| XM_006246052.2 | XP_006246114.1 | 19 | 3792 | |||
| XM_006246053.2 | XP_006246115.1 | 19 | 3578 | |||
| XM_008767606.1 | XP_008765828.1 | 19 | 4125 | |||

Cramp1l
Genomic Map