siRNA Details
| Gene Symbol: Pigq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007607.1 | NP_001007608.1 | 7 | 1356 | ||
| XM_006245942.2 | XP_006246004.1 | 7 | 1435 | |||
| XM_008767559.1 | XP_008765781.1 | 7 | 1375 | |||
| XM_008767560.1 | XP_008765782.1 | 7 | 1402 | |||
| GenBank mRNA | BC081875.1 | - | 7 | 1356 | 7107681 | |

Pigq
Genomic Map