siRNA Details
| Gene Symbol: Bod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013854.1 | NP_001013876.2 | 3 | 602 | ||
| GenBank mRNA | BC079025.1 | - | 3 | 602 | 7113945 | |
| FQ213681.1 | - | 3 | 401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bod1| Gene Symbol: Bod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013854.1 | NP_001013876.2 | 3 | 602 | ||
| GenBank mRNA | BC079025.1 | - | 3 | 602 | 7113945 | |
| FQ213681.1 | - | 3 | 401 | |||