siRNA Details
| Gene Symbol: Ttc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005529.1 | NP_001005529.1 | 5, 6 | 631 | ||
| XM_008767639.1 | XP_008765861.1 | 5, 6 | 682 | |||
| GenBank mRNA | BC082093.1 | - | 5, 6 | 631 | 7134129 | |
| FQ213119.1 | - | 5, 6 | 648 | |||

Ttc1
Genomic Map