siRNA Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105778.2 | NP_001099248.1 | 9 | 1390 | ||
| GenBank mRNA | BC168651.1 | - | 9 | 1390 | 9033412 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf145| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105778.2 | NP_001099248.1 | 9 | 1390 | ||
| GenBank mRNA | BC168651.1 | - | 9 | 1390 | 9033412 | |