siRNA Details
| Gene Symbol: Cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004200.1 | NP_001004200.1 | 7 | 787 | ||
| GenBank mRNA | BC079143.1 | - | 7 | 787 | 7128107 | |
| FQ213575.1 | - | 3 | 321 | |||
| FQ213666.1 | - | 3 | 318 | |||
| FQ228882.1 | - | 7 | 828 | |||

Cops3
Genomic Map