siRNA Details
| Gene Symbol: Cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004200.1 | NP_001004200.1 | 3 | 325 | ||
| GenBank mRNA | BC079143.1 | - | 3 | 325 | 7128107 | |
| FQ228882.1 | - | 3 | 366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cops3| Gene Symbol: Cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004200.1 | NP_001004200.1 | 3 | 325 | ||
| GenBank mRNA | BC079143.1 | - | 3 | 325 | 7128107 | |
| FQ228882.1 | - | 3 | 366 | |||