siRNA Details
| Gene Symbol: Mfap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034124.1 | NP_001029296.1 | 4 | 284 | ||
| XM_006246476.2 | XP_006246538.1 | 4 | 363 | |||
| GenBank mRNA | BC100616.1 | - | 4 | 284 | 7933452 | |
| Gene Symbol: LOC102553715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246560.2 | XP_006246622.1 | 363 | |||
| XM_006246561.2 | XP_006246623.1 | 362 | ||||

Mfap4
Genomic Map