siRNA Details
| Gene Symbol: Tekt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024739.1 | NP_001019910.1 | 2 | 617 | ||
| GenBank mRNA | BC097398.1 | - | 2 | 617 | 7453601 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tekt3| Gene Symbol: Tekt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024739.1 | NP_001019910.1 | 2 | 617 | ||
| GenBank mRNA | BC097398.1 | - | 2 | 617 | 7453601 | |