siRNA Details
| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009246.1 | NP_001009246.1 | 3 | 445 | ||
| XM_006246603.2 | XP_006246665.1 | 2 | 405 | |||
| XM_006246604.2 | XP_006246666.1 | 3 | 512 | |||
| GenBank mRNA | BC088174.1 | - | 3 | 445 | 7323419 | |
| EU037900.1 | - | 1 | 292 | |||

Adprm
Genomic Map