siRNA Details
| Gene Symbol: Dhrs7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271597.1 | NP_001258526.1 | 6 | 773 | ||
| NM_001271598.1 | NP_001258527.1 | 6 | 773 | |||
| GenBank mRNA | FQ214628.1 | - | 6 | 755 | ||
| FQ214964.1 | - | 6 | 753 | |||
| FQ215164.1 | - | 6 | 754 | |||
| FQ215595.1 | - | 6 | 754 | |||
| FQ216481.1 | - | 6 | 755 | |||
| FQ216493.1 | - | 6 | 754 | |||
| FQ216936.1 | - | 6 | 754 | |||
| FQ224852.1 | - | 6 | 754 | |||
| FQ224951.1 | - | 6 | 753 | |||

Dhrs7c
Genomic Map