siRNA Details
| Gene Symbol: Mfsd6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105787.1 | NP_001099257.1 | 1 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd6l| Gene Symbol: Mfsd6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105787.1 | NP_001099257.1 | 1 | 378 | ||