siRNA Details
| Gene Symbol: Arhgef15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105789.1 | NP_001099259.1 | 2 | 251 | ||
| XM_008767788.1 | XP_008766010.1 | 2 | 1497 | |||
| XM_008767789.1 | XP_008766011.1 | 2 | 2057 | |||
| XM_008767790.1 | XP_008766012.1 | 1 | 386 | |||
| XM_008767791.1 | XP_008766013.1 | 1 | 387 | |||
| XM_008767792.1 | XP_008766014.1 | 1 | 385 | |||
| XM_008767793.1 | XP_008766015.1 | 1 | 388 | |||

Arhgef15
Genomic Map