siRNA Details
Gene Symbol: Wrap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007610.1 | NP_001007611.1 | 3 | 640 | ||
GenBank mRNA | BC083696.1 | - | 3 | 640 | 7189640 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wrap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007610.1 | NP_001007611.1 | 3 | 640 | ||
GenBank mRNA | BC083696.1 | - | 3 | 640 | 7189640 |