siRNA Details
| Gene Symbol: Cd68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031638.1 | NP_001026808.1 | 5 | 799 | ||
| GenBank mRNA | BC098931.1 | - | 5 | 799 | 7368498 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd68| Gene Symbol: Cd68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031638.1 | NP_001026808.1 | 5 | 799 | ||
| GenBank mRNA | BC098931.1 | - | 5 | 799 | 7368498 | |