siRNA Details
| Gene Symbol: Zbtb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246843.2 | XP_006246905.1 | 2 | 227 | ||
| XM_006246844.2 | XP_006246906.1 | 1 | 51 | |||
| GenBank mRNA | CB605808.1 | - | 1 | 28 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb4| Gene Symbol: Zbtb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246843.2 | XP_006246905.1 | 2 | 227 | ||
| XM_006246844.2 | XP_006246906.1 | 1 | 51 | |||
| GenBank mRNA | CB605808.1 | - | 1 | 28 | ||