siRNA Details
| Gene Symbol: Acap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105796.2 | NP_001099266.2 | 18 | 1914 | ||
| GenBank mRNA | FQ230675.1 | - | 2 | 171 | ||
| FQ231017.1 | - | 2 | 171 | |||
| FQ231544.1 | - | 2 | 171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acap1| Gene Symbol: Acap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105796.2 | NP_001099266.2 | 18 | 1914 | ||
| GenBank mRNA | FQ230675.1 | - | 2 | 171 | ||
| FQ231017.1 | - | 2 | 171 | |||
| FQ231544.1 | - | 2 | 171 | |||