siRNA Details
| Gene Symbol: Slc16a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105797.2 | NP_001099267.2 | 6 | 1097 | ||
| XM_006246646.1 | XP_006246708.1 | 6 | 1237 | |||
| XM_006246647.2 | XP_006246709.1 | 5 | 1780 | |||
| XM_006246648.2 | XP_006246710.1 | 5 | 1082 | |||
| XM_006246649.2 | XP_006246711.1 | 4 | 801 | |||
| XM_008767805.1 | XP_008766027.1 | 5 | 1641 | |||
| GenBank mRNA | BC169003.1 | - | 6 | 1237 | 7122746 | |
| CK365106.1 | - | 2 | 708 | |||
| FQ219143.1 | - | 3 | 572 | |||

Slc16a11
Genomic Map