siRNA Details
| Gene Symbol: Alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105798.1 | NP_001099268.1 | 11 | 1497 | ||
| XM_006246659.2 | XP_006246721.1 | 10 | 1329 | |||
| GenBank mRNA | BC168184.1 | - | 11 | 1497 | 7098799 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alox12| Gene Symbol: Alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105798.1 | NP_001099268.1 | 11 | 1497 | ||
| XM_006246659.2 | XP_006246721.1 | 10 | 1329 | |||
| GenBank mRNA | BC168184.1 | - | 11 | 1497 | 7098799 | |