siRNA Details
| Gene Symbol: Zmynd15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105800.1 | NP_001099270.1 | 13 | 2549 | ||
| XM_006246661.2 | XP_006246723.1 | 12 | 2064 | |||
| XM_006246662.2 | XP_006246724.1 | 12 | 2051 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmynd15| Gene Symbol: Zmynd15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105800.1 | NP_001099270.1 | 13 | 2549 | ||
| XM_006246661.2 | XP_006246723.1 | 12 | 2064 | |||
| XM_006246662.2 | XP_006246724.1 | 12 | 2051 | |||