siRNA Details
| Gene Symbol: Foxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100648.1 | NP_001094118.1 | 5, 6 | 934 | ||
| GenBank mRNA | S80120.1 | - | 5, 6 | 934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxn1| Gene Symbol: Foxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100648.1 | NP_001094118.1 | 5, 6 | 934 | ||
| GenBank mRNA | S80120.1 | - | 5, 6 | 934 | ||