siRNA Details
| Gene Symbol: Med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135813.1 | NP_001129285.1 | 4 | 386 | ||
| GenBank mRNA | FQ211088.1 | - | 4 | 383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med31| Gene Symbol: Med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135813.1 | NP_001129285.1 | 4 | 386 | ||
| GenBank mRNA | FQ211088.1 | - | 4 | 383 | ||