siRNA Details
| Gene Symbol: Zzef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246857.2 | XP_006246919.1 | 35 | 5538 | ||
| XM_237787.8 | XP_237787.4 | 35 | 5535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zzef1| Gene Symbol: Zzef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246857.2 | XP_006246919.1 | 35 | 5538 | ||
| XM_237787.8 | XP_237787.4 | 35 | 5535 | |||