siRNA Details
| Gene Symbol: Ctns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191647.1 | NP_001178576.1 | 10 | 986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctns| Gene Symbol: Ctns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191647.1 | NP_001178576.1 | 10 | 986 | ||