siRNA Details
| Gene Symbol: Rpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047843.1 | NP_001041308.1 | 7 | 622 | ||
| XM_006246673.1 | XP_006246735.1 | 8 | 746 | |||
| XM_008767961.1 | XP_008766183.1 | 4 | 380 | |||
| GenBank mRNA | AY325234.1 | - | 7 | 622 | ||
| FQ222430.1 | - | 8 | 753 | |||

Rpa1
Genomic Map