siRNA Details
| Gene Symbol: Prpf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191590.1 | NP_001178519.1 | 17 | 2498 | ||
| XM_006246892.2 | XP_006246954.1 | 16 | 2699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prpf8| Gene Symbol: Prpf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191590.1 | NP_001178519.1 | 17 | 2498 | ||
| XM_006246892.2 | XP_006246954.1 | 16 | 2699 | |||