siRNA Details
| Gene Symbol: Blmh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034163.1 | NP_001029335.1 | 5 | 706 | ||
| GenBank mRNA | BC128701.1 | - | 5 | 591 | 7379958 | |
| CB768995.1 | - | 4 | 365 | |||
| CB772927.1 | - | 3 | 278 | |||
| CK363809.1 | - | 5 | 633 | |||
| D87336.1 | - | 5 | 609 | |||

Blmh
Genomic Map