siRNA Details
| Gene Symbol: Tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025626.1 | NP_001020797.1 | 3 | 1105 | ||
| GenBank mRNA | BC098792.1 | - | 3 | 1105 | 7375621 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tefm| Gene Symbol: Tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025626.1 | NP_001020797.1 | 3 | 1105 | ||
| GenBank mRNA | BC098792.1 | - | 3 | 1105 | 7375621 | |