siRNA Details
Gene Symbol: Tefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025626.1 | NP_001020797.1 | 3 | 849 | ||
GenBank mRNA | BC098792.1 | - | 3 | 849 | 7375621 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025626.1 | NP_001020797.1 | 3 | 849 | ||
GenBank mRNA | BC098792.1 | - | 3 | 849 | 7375621 |