siRNA Details
| Gene Symbol: Slfn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750873.3 | XP_003750921.1 | 2 | 833 | ||
| GenBank mRNA | AY539914.1 | - | 951 | |||
| Gene Symbol: LOC103689937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727784.4 | XP_002727830.2 | 2 | 833 | ||

Slfn14
Genomic Map