siRNA Details
Gene Symbol: Ppm1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105825.2 | NP_001099295.2 | 2 | 750 | ||
GenBank mRNA | BC162058.1 | - | 2 | 750 | 7115026 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppm1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105825.2 | NP_001099295.2 | 2 | 750 | ||
GenBank mRNA | BC162058.1 | - | 2 | 750 | 7115026 |