siRNA Details
Gene Symbol: Supt4h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105828.2 | NP_001099298.2 | 1, 2 | 81 | ||
GenBank mRNA | BC168730.1 | - | 1, 2 | 81 | 6887426 | |
FQ213593.1 | - | 1, 2 | 89 | |||
FQ217757.1 | - | 1, 2 | 88 | |||
FQ224188.1 | - | 1, 2 | 77 | |||
FQ228457.1 | - | 1, 2 | 75 |
