siRNA Details
| Gene Symbol: Supt4h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105828.2 | NP_001099298.2 | 2 | 130 | ||
| GenBank mRNA | BC168730.1 | - | 2 | 130 | 6887426 | |
| FQ213593.1 | - | 2 | 138 | |||
| FQ217757.1 | - | 2 | 137 | |||
| FQ224188.1 | - | 2 | 126 | |||

Supt4h1
Genomic Map