siRNA Details
| Gene Symbol: Lpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105829.1 | NP_001099299.1 | 5 | 736 | ||
| XM_006247085.2 | XP_006247147.1 | 5 | 809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpo| Gene Symbol: Lpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105829.1 | NP_001099299.1 | 5 | 736 | ||
| XM_006247085.2 | XP_006247147.1 | 5 | 809 | |||