siRNA Details
| Gene Symbol: Vezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768166.1 | XP_008766388.1 | 2 | 475 | ||
| XM_008768167.1 | XP_008766389.1 | 2 | 475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vezf1| Gene Symbol: Vezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768166.1 | XP_008766388.1 | 2 | 475 | ||
| XM_008768167.1 | XP_008766389.1 | 2 | 475 | |||