siRNA Details
| Gene Symbol: Slc35b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199081.1 | NP_954512.1 | 7 | 793 | ||
| GenBank mRNA | AY349135.1 | - | 7 | 793 | ||
| BC085347.1 | - | 7 | 828 | 7305181 | ||
| D87991.1 | - | 7 | 786 | |||
| FQ233464.1 | - | 7 | 816 | |||

Slc35b1
Genomic Map