siRNA Details
| Gene Symbol: Spop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100496.1 | NP_001093966.1 | 6 | 758 | ||
| XM_006247180.2 | XP_006247242.1 | 7 | 1562 | |||
| XM_006247181.2 | XP_006247243.1 | 8 | 869 | |||
| XM_006247182.2 | XP_006247244.1 | 7 | 818 | |||
| XM_008767968.1 | XP_008766190.1 | 7 | 914 | |||
| XM_008767969.1 | XP_008766191.1 | 8 | 836 | |||
| GenBank mRNA | BC091435.1 | - | 4 | 484 | ||
| BC166743.1 | - | 6 | 758 | 7938440 | ||
| FQ217100.1 | - | 8 | 1066 | |||

Spop
Genomic Map