siRNA Details
| Gene Symbol: Lrrc46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004201.1 | NP_001004201.1 | 1, 2 | 248 | ||
| XM_006247268.2 | XP_006247330.1 | 1, 2 | 78 | |||
| GenBank mRNA | BC079258.1 | - | 1, 2 | 248 | 7133394 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc46| Gene Symbol: Lrrc46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004201.1 | NP_001004201.1 | 1, 2 | 248 | ||
| XM_006247268.2 | XP_006247330.1 | 1, 2 | 78 | |||
| GenBank mRNA | BC079258.1 | - | 1, 2 | 248 | 7133394 | |