siRNA Details
| Gene Symbol: Gpr179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768197.1 | XP_008766419.1 | 11 | 6862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr179| Gene Symbol: Gpr179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768197.1 | XP_008766419.1 | 11 | 6862 | ||