siRNA Details
| Gene Symbol: Psmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008281.1 | NP_001008282.1 | 2 | 510 | ||
| XM_006247272.1 | XP_006247334.1 | 2 | 725 | |||
| GenBank mRNA | BC085881.1 | - | 2 | 510 | 7191926 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd3| Gene Symbol: Psmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008281.1 | NP_001008282.1 | 2 | 510 | ||
| XM_006247272.1 | XP_006247334.1 | 2 | 725 | |||
| GenBank mRNA | BC085881.1 | - | 2 | 510 | 7191926 | |