siRNA Details
| Gene Symbol: Ifi35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009625.1 | NP_001009625.1 | 5 | 610 | ||
| GenBank mRNA | BC088222.1 | - | 5 | 610 | 7321956 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi35| Gene Symbol: Ifi35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009625.1 | NP_001009625.1 | 5 | 610 | ||
| GenBank mRNA | BC088222.1 | - | 5 | 610 | 7321956 | |