siRNA Details
Gene Symbol: Ifi35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009625.1 | NP_001009625.1 | 7 | 879 | ||
GenBank mRNA | BC088222.1 | - | 7 | 879 | 7321956 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifi35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009625.1 | NP_001009625.1 | 7 | 879 | ||
GenBank mRNA | BC088222.1 | - | 7 | 879 | 7321956 |