siRNA Details
| Gene Symbol: FAM187A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013861.1 | NP_001013883.1 | 1 | 1410 | ||
| GenBank mRNA | BC079299.1 | - | 1 | 1410 | 7134519 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM187A| Gene Symbol: FAM187A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013861.1 | NP_001013883.1 | 1 | 1410 | ||
| GenBank mRNA | BC079299.1 | - | 1 | 1410 | 7134519 | |