siRNA Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105845.2 | NP_001099315.2 | 4, 5 | 768 | ||
| XM_006247494.2 | XP_006247556.1 | 4, 5 | 809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcd3| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105845.2 | NP_001099315.2 | 4, 5 | 768 | ||
| XM_006247494.2 | XP_006247556.1 | 4, 5 | 809 | |||