siRNA Details
| Gene Symbol: Spata32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005531.1 | NP_001005531.1 | 4 | 712 | ||
| GenBank mRNA | AF473841.1 | - | 1 | 494 | ||
| BC082079.1 | - | 4 | 712 | 7133280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata32| Gene Symbol: Spata32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005531.1 | NP_001005531.1 | 4 | 712 | ||
| GenBank mRNA | AF473841.1 | - | 1 | 494 | ||
| BC082079.1 | - | 4 | 712 | 7133280 | ||