siRNA Details
| Gene Symbol: Ddx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007613.1 | NP_001007614.1 | 12 | 1413 | ||
| XM_008768312.1 | XP_008766534.1 | 11 | 1402 | |||
| XM_008768313.1 | XP_008766535.1 | 11 | 1325 | |||
| GenBank mRNA | BC079036.1 | - | 12 | 1413 | 7114313 | |
| FJ168767.1 | - | 11 | 1244 | |||

Ddx5
Genomic Map