siRNA Details
| Gene Symbol: Cep95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013862.1 | NP_001013884.1 | 9 | 1316 | ||
| GenBank mRNA | BC083896.1 | - | 9 | 1316 | 7133061 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep95| Gene Symbol: Cep95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013862.1 | NP_001013884.1 | 9 | 1316 | ||
| GenBank mRNA | BC083896.1 | - | 9 | 1316 | 7133061 | |