siRNA Details
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013862.1 | NP_001013884.1 | 15 | 2051 | ||
GenBank mRNA | BC083896.1 | - | 15 | 2051 | 7133061 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013862.1 | NP_001013884.1 | 15 | 2051 | ||
GenBank mRNA | BC083896.1 | - | 15 | 2051 | 7133061 |