siRNA Details
| Gene Symbol: Slc39a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013042.1 | NP_001013060.1 | 5, 6 | 530 | ||
| XM_006247665.2 | XP_006247727.1 | 5, 6 | 828 | |||
| GenBank mRNA | BC062054.1 | - | 5, 6 | 530 | 5598901 | |

Slc39a11
Genomic Map