siRNA Details
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_221081.8 | XP_221081.7 | 9 | 1213 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_221081.8 | XP_221081.7 | 9 | 1213 |